Learn More
Medchemexpress LLC Nafcillin sodium monohydrate | 7177-50-6 | 98.1% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0555A50MG
Encompass_Preferred
Nafcillin sodium monohydrate is an antibiotic that acts as a reversible inhibitor of β-lactamase. It is suitable for the research of staphylococcal infections and is for research use only.
  • Reversible inhibitor of β-lactamase.
  • Suitable for research of staphylococcal infections.
  • Exhibits bactericidal activity against methicillin-susceptible *Staphylococcus aureus* (MSSA) and methicillin-resistant *S. aureus* (MRSA) in mice.
  • Protected 100% of healthy mice with MSSA infections.
Catalog No. 50-004-07845