Learn More
Abcam Naftopidil hydrochloride, alpha1 Adrenoceptor antagonist, 50MG
Supplier:  Abcam AB12075650MG
Encompass_Preferred
MW 428.96 g/mol, Purity >98%. α₁ Adrenoceptor antagonist. pKi values are 3.7, 20 and 1.2 for α₁a, α₁b and α₁d, respectively. Orally active. Implicated in benign prostate hyperplasia and bladder obstruction. Antihypertensive.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7305