Learn More
Selleck Chemical LLC Naftopidil 25mg 57149-07-2 KT-611
SDP
Supplier:  Selleck Chemical LLC S212625MG
Encompass_Preferred
Naftopidil (KT-611) is a selective ?1-adrenergic receptor antagonist with Ki of 3.7 nM, 20 nM, and 1.2 nM for ?1a, ?1b, and ?1d, respectively, used for the treatment of benign prostatic hyperplasia. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-664-3