Learn More
ATCC Naganishia albida (Saito) Liu et al. freeze-dried culture, strain 378 - ATCC 26803
Supplier:  ATCC 26803
Encompass
Naganishia albida (Saito) Liu et al. (strain designation 378) is supplied as a freeze-dried ampoule containing viable yeast cells suspended in cryoprotectant. The culture is intended for laboratory research, reference, and quality-control applications requiring a well-characterized yeast strain.
  • Freeze-dried ampoule containing viable cells.
  • Viable cells may include spores and mycelia.
  • Recommended storage 2 °C to 8 °C.
  • Strain designation 378.
  • Synonyms include Cryptococcus albidus, Torula albida, Torulopsis albida, and Rhodotorula albida.
  • Suitable for research, reference, and quality-control use.
Catalog No. NC3684550