Learn More
Medchemexpress LLC HY-103006 10mg Medchemexpress, NAI-N3 CAS:1612756-29-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10300610MG
Encompass_Preferred
Medchemexpress, HY-103006 10mg NAI-N3 CAS:1612756-29-2 NAI-N3 is a RNA acylation reagent that enables RNA purification. NAI-N3 is a dual-function SHAPE (selective 2-hydroxyl acylation and profiling experiment) probe (RNA structure probe and enrichment). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8721