Learn More
Hach Company Nalgene Laboratory Pen
Supplier:  Hach Company 2092000
Encompass
Use this felt-tip pen for permanent marking with black ink on glass and plastic items Ink dries quickly Markings on glass or plastic are not affected by repeated autoclaving
Catalog No. 50-002-57044