Learn More
Medchemexpress LLC Nalidixic acid | 389-08-2 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB039810MM/1ML
Encompass_Preferred
Nalidixic acid is a quinolone antibiotic effective against both gram-positive and gram-negative bacteria. It acts in a bacteriostatic manner at lower concentrations and is bactericidal at higher concentrations. This antibiotic functions by inhibiting a subunit of DNA gyrase and topoisomerase IV, thereby reversibly blocking DNA replication in susceptible bacteria.
  • Effective against gram-positive and gram-negative bacteria
  • Inhibits DNA gyrase and topoisomerase IV
  • Reversibly blocks DNA replication
  • Active against *Escherichia coli*, *Pasteurella spp.*, and *Klebsiella pneumoniae*
  • Pronounced in vivo activity against Gram-negative bacteria
Catalog No. 50-004-06655