Learn More
Cayman Chemical Naloxon benzoylhydrazon 5mg
SDP
Supplier:  Cayman Chemical 398275 MG
Encompass_Preferred
A μ-, κ1-, and δ-opioid receptor antagonist (pA2s = 8.82, 7.45, and 7.76, respectively, for the mouse receptors); a κ3-opioid receptor agonist (IC50 = 0.7 nM for the guinea pig receptor); inhibits increases in the latency to tail withdraw induced by DAMGO in a hotplate assay in mice at 1 UG/animal, i.c.v.; increases food intake in rats at 0.1, 1, and 20 UG/animal, i.c.v.
Catalog No. 50-294-2093