Learn More
Medchemexpress LLC Naloxone | 465-65-6 | 99.7% | 327.37 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY17417A50MG
Encompass_Preferred
Naloxone is an antagonist of the Opioid receptor. It alleviates opioid-overdose-induced respiratory depression and may cause pulmonary edema and cardiac arrhythmias. It is for research use only and not sold to patients.
  • Alleviates opioid-overdose-induced respiratory depression
  • May cause pulmonary edema and cardiac arrhythmias
  • For research use only
Catalog No. 50-003-85646