Learn More
Medchemexpress LLC Naloxone benzoylhydrazone | 119630-94-3 | MFCD05662358 | 98.4% | 445.51 | C26H27N3O4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1077431MG
Encompass_Preferred
Naloxone benzoylhydrazone is a naloxone-derived small molecule described as a mixed agonist/antagonist with prototypic κ3-opioid receptor agonist activity; it is used in pharmacology research to investigate opioid receptor signaling and analgesic responses.
  • Prototypic κ3-opioid receptor agonist activity.
  • Partial agonist at μ and δ opioid receptors.
  • Antagonist activity at opioid-like NOP receptors.
  • High reported purity (98.36%).
  • Solid form with defined storage conditions for stability.
Catalog No. 50-002-41184