Learn More
Medchemexpress LLC Naloxone hydrochloride | 357-08-4 | 100.0% | 363.84 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY17417100MG
Encompass_Preferred
Naloxone hydrochloride is an antagonist of the Opioid receptor. It alleviates respiratory depression induced by opioid overdose, though it may also cause pulmonary edema and cardiac arrhythmias. It is intended for research use only and not for sale to patients.
  • Opioid receptor antagonist
  • Alleviates opioid-overdose-induced respiratory depression
  • Data sheet, COA, SDS, and handling instructions provided
  • Soluble in H2O (62.5 mg/mL) and DMSO (≥ 30 mg/mL)
Catalog No. 50-003-84025