Learn More
Medchemexpress LLC Naloxone hydrochloride | 357-08-4 | 99.9% | 363.84 | 25MG

Supplier: Medchemexpress LLC 25MGHY17417A
Naloxone hydrochloride is an antagonist of the Opioid receptor. It alleviates opioid-overdose-induced respiratory depression and is intended for research use only, not for human patient use.
- Antagonist of the Opioid receptor
- Alleviates opioid-overdose-induced respiratory depression
- Soluble in water at 62.5 mg/mL (171.78 mM) with ultrasonic assistance
- Soluble in DMSO at ≥ 30 mg/mL (82.45 mM)
- Recommended storage at 4°C, sealed and away from moisture
- In solvent, store at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.