Learn More
Medchemexpress LLC Naloxone hydrochloride | 357-08-4 | 99.96% | 363.84 | 100MG
SDP
Supplier:  Medchemexpress LLC HY17417A100MG
Encompass_Preferred
Naloxone hydrochloride is an opioid receptor antagonist that alleviates opioid-overdose-induced respiratory depression. It is intended for research use only and not for patient administration. The compound may cause pulmonary edema and cardiac arrhythmias. It is soluble in both water and DMSO.
  • Acts as an antagonist of the opioid receptor
  • Alleviates respiratory depression caused by opioid overdose
  • Soluble in water and DMSO for various research applications
  • Available in solid, white to off-white appearance
Catalog No. 50-003-83994