Learn More
Medchemexpress LLC Naluzotan | 740873-06-7 | 98.05% | 450.64 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY148481MG
Encompass_Preferred
Naluzotan is a novel, potent, and selective amidosulfonamide 5-HT1A agonist with IC50 and Ki of approximately 20 nM and 5.1 nM, respectively. It is used for the treatment of anxiety and depression. It also acts as a weak hERG K+ channel blocker, with an IC50 of 3800 nM.
  • Novel, potent, and selective amidosulfonamide 5-HT1A agonist
  • Used for the treatment of anxiety and depression
  • Acts as a weak hERG K+ channel blocker
  • Exhibits full agonism in in vitro functional assays
  • Shows significant affinity for the guinea pig sigma receptor
  • Does not inhibit major cytochrome P450 isoforms
  • Demonstrates significant brain penetration
Catalog No. 50-003-61319