Learn More
TARGETMOL CHEMICALS INC NAMIROTENE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T335845MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Namirotene (CBS 211A) is a protein biosynthesis inhibitor a synthetic retinoic acid analog that is used in the study of corneal diseases corneal ulcers and dry eye syndrome. purity: 99%
Catalog No. 50-258-0299