Learn More
AdipoGen anti-Nampt (Visfatin/PBEF), mAb (OMNI379) (50 µg)
SDP
Supplier:  AdipoGen AG20A0034C050
Encompass_Preferred
Monoclonal Antibody. Recognizes human, mouse and rat Nampt. Isotype: Mouse IgG2akappa. Clone: OMNI379. Applications: ELISA, FACS, ICC, IHC, IP, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. Nicotinamide phosphoribosyltransferase (Nampt; pre-B cell colony-enhancing factor; PBEF; Visfatin) is an 52kDa adipokine secreted by adipose tissue and involved in the biosynthesis of nicotinamide adenine dinucleotide (NAD+). Two forms of Nampt exist, an intracellular form (iNampt) and an extracellular form (eNampt). While the function of iNampt as an essential and rate-limiting NAD+ biosynthetic enzyme is well established, the physiological role of eNampt is still a matter of debate. Nampt has various functions, including the promotion of vascular smooth muscle cell maturation and inhibition of neutrophil apoptosis. It activates insulin receptor and has insulin-mimetic effects, lowering blood glucose and improving insulin sensitivity.
Catalog No. 50-205-2821