Learn More
Enzo Life Sciences Nampt (Visfatin/PBEF) (human), (recombinant) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2650100
Encompass_Preferred
Alternative name: Pre B cell colony enhancing factor 1, Nicotinamide phosphoribosyltransferase. Purity: ≥90% (SDS-PAGE). Formulation: Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 0.1% Trifluoroacetic Acid (TFA). Source: Produced in E. coli. Contains 466 amino acids. Long Term Storage: -80°C.
Catalog No. 50-201-4081