Learn More
Medchemexpress LLC Nan-190 (hydrobromide) | 115338-32-4 | 99.1% | 474.39 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY19818A5MG
Encompass_Preferred
NAN-190 hydrobromide is a serotonin receptor 5-HT antagonist and a selective antagonist of 5-HT1A. This compound is intended for research use only.
  • Serotonin receptor 5-HT antagonist
  • Selective antagonist of 5-HT1A
  • Targets 5-HT1A receptor and 5-HT1 receptor
  • Involved in GPCR/G protein and neuronal signaling pathways
  • Reverses the catalepsy-improving effect of fluoxetine in 6-OHDA lesioned rats
Catalog No. 50-004-01680