Learn More
Abcam NAN-190 hydrobromide, 5-HT1A antagonist, 250MG
Supplier:  Abcam AB120543250MG
Encompass_Preferred
MW 474.4 g/mol, Purity >99%. Potent 5-HT₁A receptor antagonist (Ki = 6.8 nM). Also displays activity at α₂ adrenoceptors. Displays anxiogenic and anxiolytic effects. Additionally, potentiates circadian rhythm response to light.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7505