Learn More
Apexbio Technology LLC Nanchangmycin 65101-87-3 5mg
SDP
Supplier:  Apexbio Technology LLC B21615
Encompass_Preferred
Nanchangmycin (65101-87-3) is a polyether ionophore antibiotic designed to facilitate the transport of monovalent cations across lipid bilayers thereby disrupting ion transport mechanisms in cellular membranes Nanchangmycin exerts its biological activity primarily through acting as an ionophore that compromises cellular ionic gradients In in vitro studies using nematode and insect models Nanchangmycin demonstrates potent inhibitory activity on nematode growth and development with reported IC50 values ranging from low nanomolar to micromolar concentrations depending on species and assay conditions Based on these pharmacological properties Nanchangmycin holds research potential in studies of membrane function ion transport mechanisms host-pathogen interactions and pesticide target validation
Catalog No. 50-000-92459