Learn More
Optimize Technologies EXP™ Nano 6-40 Hex Head Nut & Nano Titanium Hybrid Ferrule (15-19-04248, 15-19-04242)
SDP
Supplier:  Optimize Technologies 151904248
Encompass_Preferred
The EXP™ Fitting System is the premier adjustable nut and ferrule compression fitting for extreme high-pressure connections between 1/32” or 1/16” tubing and any 6-32, 6-40 or 10-32 port. EXP Fittings are simple and versatile. The patented Titanium Hybrid Ferrule integrates the resilience of titanium with the proven sealing ability of PEEK in a superb one-piece design. The Hybrid Ferrule provides a perfect seal with every connection. The swage can be released without tools to adjust to the different port depths of various hardware. This adjustability provides value through repeated zero-dead-volume connections with any port configuration.
Catalog No. 50-200-6557