Learn More
CTL Scientific Supply Corp NANOCOLOR COD 60 in Saltwater, contains 20 tube tests
SDP
Supplier:  CTL Scientific Supply Corp 985020
Encompass_Preferred
Tube test for the determination of COD in saltwater. This test kit is suitable for chloride-containing wastewater, surface water and seawater with chloride concentrations of 2–18 g/L Cl-. Precise rapid tests for all kind of water and waste water samples according to ISO 15705. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. *This item is hazardous and cannot ship Parcel Post. It is required to ship UPS Ground* REFRIGERATED ITEMManufactured by Macherey-Nagel
Catalog No. NC2958210