Learn More
Spherotech NANO FLRSCNT SZ STND YLLW
Supplier:  Spherotech NFPPS01525
Encompass_Preferred
Nano Fluorescent Particle, Flow Cytometry Grade, Yellow, 1E6/mL, 0.05-0.15 um, 5 mL
Catalog No. NC0650348