Learn More
Molecular Devices Molecular Devices Detection Reagent, Human IgG (H+L), Fluorescein Conjugate, BSA-Free
Supplier: Molecular Devices K8202
CloneDetect Human IgG (H+L) Specific is a fluorescein (FITC)-conjugated, affinity-purified polyclonal detection reagent raised in sheep for sensitive detection of human IgG heavy and light chains. The reagent is supplied sterile and formulated without bovine serum albumin or azide for use in clone screening and immunoassays.
- Host species: Sheep.
- Target specificity: Human IgG (H+L).
- Conjugate: Fluorescein (FITC).
- Formulation: Phosphate-buffered saline, pH 7.2; BSA-free, azide-free.
- Sterility: Sterile, 0.2 μm filtered.
- Concentration: 10,000 U/mL.
- Volume: 1.0 mL.
- Total protein: 0.75 mg.
- Storage: 2-8 °C.
- Intended use: For research use only.
Description
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.