Learn More
CTL Scientific Supply Corp NANOCOLOR ortho and total Phosphate LR 1, contains 20 tube tests
SDP
Supplier:  CTL Scientific Supply Corp 985095
Encompass_Preferred
Tube test for the determination of ortho- and total Phosphate. Precise rapid tests for all kind of water and waste water samples. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: Phosphate (ortho) - 0.05–0.50 mg/L P , Phosphate (ortho) - 0.2–1.5 mg/L PO₄³⁻ , Phosphate (total) - 0.05–0.50 mg/L P , Phosphate (total) - 0.2–1.5 mg/L PO₄³⁻Manufactured by Macherey-Nagel
Catalog No. NC2958208