Learn More
CTL Scientific Supply Corp NANOCOLOR total Kjeldahl Nitrogen TKN 16, contains 20 tube tests
SDP
Supplier:  CTL Scientific Supply Corp 985067
Encompass_Preferred
Tube test for the determination of total Kjeldahl Nitrogen (TKN). Precise rapid tests for all kind of water and waste water samples. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: Kjeldahl-Nitrogen - 1.00–16.0 mg/L TKN *This item is hazardous and cannot ship Parcel Post. It is required to ship UPS Ground*Manufactured by Macherey-Nagel
Catalog No. NC2958190