Learn More
CTL Scientific Supply Corp NANOCOLOR COD 1500 Hg-free, contains 20 tube tests
SDP
Supplier:  CTL Scientific Supply Corp 963029
Encompass_Preferred
Tube test for the determination of COD. Precise rapid tests for all kind of water and waste water samples. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: COD - 100–1500 mg/L O₂ *This item is hazardous and cannot ship Parcel Post. It is required to ship UPS Ground*Manufactured by Macherey-Nagel
Catalog No. NC2958213