Learn More
CTL Scientific Supply Corp NANOCOLOR Ammonium 100, contains 20 tube tests
SDP
Supplier:  CTL Scientific Supply Corp 985008
Encompass_Preferred
Tube test for the determination of Ammonium. Precise rapid tests for all kind of water and waste water samples. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: Ammonium - 4–80 mg/L NH₄-N, Ammonium - 5–100 mg/L NH₄⁺Manufactured by Macherey-Nagel
Catalog No. NC2958045