Learn More
CTL Scientific Supply Corp NANOCOLOR Nitrate 250, contains 20 tube tests
SDP
Supplier:  CTL Scientific Supply Corp 985066
Encompass_Preferred
Tube test for the determination of Nitrate. Precise rapid tests for all kind of water and waste water samples. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: Nitrate-4–60 mg/L NO₃-N, Nitrate - 20–250 mg/L NO₃⁻ *This item is hazardous and cannot ship Parcel Post. It is required to ship UPS Ground*Manufactured by Macherey-Nagel
Catalog No. NC2958199