Learn More
CTL Scientific Supply Corp NANOCOLOR Nitrite 2, contains 20 tube tests
SDP
Supplier:  CTL Scientific Supply Corp 985068
Encompass_Preferred
Tube test for the determination of Nitrite. Precise rapid tests for all kind of water and waste water samples. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: Nitrite - 0.003–0.460 mg/L NO₂-N, Nitrite - 0.02–1.50 mg/L NO₂⁻Manufactured by Macherey-Nagel
Catalog No. NC2958172