Learn More
CTL Scientific Supply Corp NANOCOLOR Nitrite 4, contains 20 tube tests
SDP
Supplier:  CTL Scientific Supply Corp 985069
Encompass_Preferred
Tube test for the determination of Nitrite. Precise rapid tests for all kind of water and waste water samples. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: Nitrite -0.1–4.0 mg/L NO₂-N, Nitrite - 0.3–13.0 mg/L NO₂⁻Manufactured by Macherey-Nagel
Catalog No. NC2958174