Learn More
CTL Scientific Supply Corp NANOCOLOR Oxygen 12, contains 22 tube tests
SDP
Supplier:  CTL Scientific Supply Corp 985082
Encompass_Preferred
Tube test for the determination of Oxygen. Precise rapid tests for all kind of water and waste water samples. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: Oxygen - 0.5–12.0 mg/L O₂ *This item is hazardous and cannot ship Parcel Post. It is required to ship UPS Ground*Manufactured by Macherey-Nagel
Catalog No. NC2958226