Learn More
CTL Scientific Supply Corp NANOCOLOR Starch 100, contains 19 tube tests
SDP
Supplier:  CTL Scientific Supply Corp 985085
Encompass_Preferred
Tube test for the determination of Starch. Precise rapid tests for all kind of water and waste water samples. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: Starch - 5–100 mg/L StarchManufactured by Macherey-Nagel
Catalog No. NC2958224