Learn More
CTL Scientific Supply Corp NANOCOLOR TTC / Sludge acitivity 150, contains 20 tube tests
SDP
Supplier:  CTL Scientific Supply Corp 985890
Encompass_Preferred
Tube test for the determination of TTC. Precise rapid tests for all kind of water and waste water analysis. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: TTC - 0.050–2.300 A , TTC - 5–150 µg TPF *This item is hazardous and cannot ship Parcel Post. It is required to ship UPS Ground* REFRIGERATED ITEMManufactured by Macherey-Nagel
Catalog No. NC2958216