Learn More
Waters Corp Column, BEH130 C18, 3.5 μm, 75 μm x 150 mm
Supplier:  Waters Corp 186010258
Encompass_Preferred
These nanoEase M/Z BEH130 C18 columns deliver optimal performance for nano- to micro-scale liquid chromatographic separations. Ideal for biomarker discovery and protein/peptide identification, they operate up to 15,000 psi (1,034 Bar) while effectively managing pressure fluctuations for superior longevity.
  • Designed for optimal nano- to micro-scale liquid chromatographic separation.
  • Supports biomarker discovery and protein/peptide identification and characterization.
  • Withstands up to 15,000 psi (1,034 Bar) operating pressure.
  • Ensures superior longevity by handling pressure fluctuations effectively.
  • Equipped with a ZenFit connector for easy, fingertight, tool-free, and leak-free liquid connections.
  • ZenFit fittings are compatible with V-Detail ports and Rheodyne/Vici compatible valves.
  • Compatible with ACQUITY UPLC M-Class Systems.
Catalog No. 50-239-2566