Learn More
Waters Corp nanoEase M/Z HSS T3 Column, 100Å, 1.8 μm
Supplier: Waters Corp 186008816
These columns are engineered for optimal performance in nano- to micro-scale liquid chromatographic separation, delivering high sensitivity, resolution, and reproducibility for advanced biomarker and protein analysis. They feature an ultra-low dispersion design and ZenFit connectors.
- Optimal performance in nano- to micro-scale liquid chromatographic separation.
- Provides high sensitivity, resolution, and reproducibility for advanced biomarker and protein analysis.
- Ultra-low dispersion design.
- Withstands 15000 psi (1034 Bar) operating pressure and manages pressure fluctuations.
- Utilizes HSS T3, 100Å pore, 1.8 μm particles.
- Suitable for silica-based selectivities and enhanced retention of hydrophilic peptides.
- Equipped with ZenFit connectors for easy, fingertight, tool-free liquid connections.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.