Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Column, 1.8 μm Particle, 100 Å Pore

Supplier:  Waters Corp 186008820

Encompass_Preferred

The nanoEase M/Z HSS T3 Column is designed for optimal performance in nano- to micro-scale liquid chromatographic separations, offering high sensitivity, resolution, and reproducibility. It features HSS T3, 100Å pore, 1.8 μm particles, and a ZenFit connector for leak-free liquid connections.

  • Optimized for nano- to micro-scale liquid chromatographic separations.
  • Provides high sensitivity, resolution, and reproducibility for biomarker discovery and protein identification.
  • Features ultra-low dispersion design for 300 μm ID columns.
  • Withstands 15,000 psi operating pressure; offers superior longevity.
  • ZenFit connector provides easy, fingertight, tool-free, leak-free liquid connections.
  • Compatible with V-Detail ports and Rheodyne/Vici compatible valves.
  • Manufactured under strict quality assurance standards.

Catalog No. 50-239-2047


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.