Learn More
Waters Corp Peptide BEH C18 Column, 300Å, 1.7 μm, 75 μm x 100 mm
Supplier:  Waters Corp 186008798
Encompass_Preferred
These columns are designed for optimal performance in nano- to micro-scale liquid chromatographic separation, offering high sensitivity, resolution, and reproducibility for biomarker discovery and protein identification. They withstand 15000 psi (1034 Bar) operating pressure, contributing to superior longevity.
  • Features BEH Technology with 300Å pore, C18, 1.7 μm particles.
  • Wide-pore (300Å) BEH particle offers wide pH range, superior low pH stability, and ultra-low column bleed.
  • Ideal for assaying large peptides in proteomics, protein characterization, and peptide synthesis.
  • QC tested for peptide separation standards using tryptic digest of cytochrome c.
  • ZenFit connector provides easy, fingertight, tool-free, reproducible, and leak-free liquid connections.
  • Fittings compatible with V-Detail ports and Rheodyne/Vici compatible valves.
Catalog No. 50-239-1099