Learn More
Waters Corp nanoEase M/Z Peptide BEH C18 Column, 300Å, 1.7 μm, 300 μm x 100 mm
Supplier:  Waters Corp 186009264
Encompass_Preferred
The nanoEase M/Z Peptide BEH C18 Column is engineered for optimal performance in nano- to micro-scale liquid chromatographic separations. It offers superior sensitivity, resolution, and reproducibility for biomarker discovery and protein identification. Featuring an ultra-low dispersion design, it withstands 15kpsi operating pressure and incorporates patented BEH Technology with 300Å pore, C18, 1.7 μm particles, ensuring a wide usable pH range and excellent low pH stability.
  • Optimal for nano- to micro-scale liquid chromatographic separation.
  • Provides high sensitivity, resolution, and reproducibility for biomarker discovery.
  • Withstands 15kpsi operating pressure.
  • Features patented BEH Technology with 300Å pore, C18, 1.7 μm particles.
  • Each batch QC tested using a gradient separation of a tryptic digest of cytochrome c.
Catalog No. 50-239-0458