Learn More
Waters Corp Waters Column, C18, 1.7 μm, 130 Å, 75 μm x 250 mm
Supplier:  Waters Corp 186008810
Encompass_Preferred
  • Optimal performance for nano- to micro-scale LC separations with high sensitivity, resolution, and reproducibility for biomarker and protein analysis.
  • Withstands 15,000 psi (1,000 Bar) operating pressure, ensuring superior longevity by managing injection pressure fluctuations.
  • Equipped with an easy, fingertight, tool-free, and leak-free connector compatible with V-Detail ports and Rheodyne/Vici valves.
  • Features CSH Technology with 130Å pore, C18, 1.7 μm particles.
  • CSH particles provide excellent peak shape for peptides with FA- or TFA-containing mobile phases.
  • Ideal for LC or LC-MS applications due to low dependence on strong ion pairing agents.
  • Each batch QC tested with a tryptic digest of cytochrome c gradient separation.
  • Functions effectively across the 1 to 10 pH range.
Catalog No. 50-146-1975