Learn More
Waters Corp Waters nanoEase M/Z Peptide Column, C18, 300Å, 1.7 μm
Supplier: Waters Corp 186009265
nanoEase M/Z Peptide BEH C18 Columns are engineered for optimal performance in nano- to micro-scale liquid chromatographic separation. They provide high sensitivity, resolution, and reproducibility for biomarker discovery, protein identification, and characterization. These columns endure 15000 psi (1034 Bar) operating pressure and effectively manage pressure fluctuations, ensuring extended longevity.
- Features patented BEH Technology with 300Å pore, C18, 1.7 μm particles.
- Designed for ultra-low dispersion in 300μm ID columns.
- Offers wide pH range, low pH stability, and minimal column bleed for large peptides.
- Includes ZenFit Connectors to simplify critical UPLC fluid connections.
- Manufactured in an ISO 9001-certified facility with individual eCord™ Intelligent Chip documentation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.