Learn More
LUNA NANOTECH INC Nanoelute Genomic Dna Extraction Kit, 250 Preps
Supplier:  LUNA NANOTECH INC NK361250
Encompass_Preferred
PuroSPIN™ NanoElute™ Genomic DNA Purification Kit, 250 Preps
The PuroSPIN™ NanoElute™ Genomic DNA Purification Kit is designed to purify genomic DNA from different biological samples such as bacteria, yeast, mammalian cells, blood, and animal tissue. Our protocol allows genomic DNA to be released from cells and purified from protein, salt, and other contaminants. Our kit uses highly efficient RNase and optimized buffer formulations to ensure efficient removal of contaminating RNA.
In comparison to our standard PuroSPIN™ Genomic DNA Purification Kit, the NanoElute version of the kit is supplemented with Nano silica spin columns instead of mini silica columns. Nano columns provide the highest possible concentration of purified genomic DNA as the elution volume needed is as little as 6 μL.
The PuroSPIN™ NanoElute™ Genomic DNA Purification Kit is particulary superior to the standard kit when the amount of th
Catalog No. 50-256-6930