Learn More
ABclonal Technology Nanog Rabbit mAb
SDP
Supplier:  ABclonal Technology A25887100UL
Encompass_Preferred
The protein encoded by this gene is a DNA binding homeobox transcription factor involved in embryonic stem (ES) cell proliferation renewal and pluripotency The encoded protein can block ES cell differentiation and can also autorepress its own expression in differentiating cells Several transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-12305