Learn More
Bioworld Nanogold*- mouse (NMI) AB, 1 mL
Supplier:  Bioworld 203300072
Encompass_Preferred
Nanogold* shines when it comes to EM. It is the new gold standard. Nanogold* reagents diffuse easily tens of microns into tissues and into permeabilized cells and cell nuclei, to where most colloidal gold probes have difficulty in reaching. Smallest comme
Catalog No. 50-198-349