Learn More
Cytiva (Formerly Pall Lab) Nanosep Centrifugal Devices with wwPTFE membrane, 0.45 m, clear(100/pkg)
Supplier:  Cytiva ODPTFE04C34
Encompass
Simple, reliable processing samples of 50 to 500 l.Available with low protein-binding Omega, Bio-Inert, wwPTFE membranes, and nucleic acid binding glass fiber membranesOmega modified PES membranes come in a wide range of MWCOs, color-coded for easy identification and result in typical recoveries are >90%Bio-Inert modified nylon membranes exhibit 25-50% lower binding than traditional nylon membraneswwPTFE is an all-purpose, hydrophilic membrane that offers low protein binding and low levels of UV-absorbing extractablesNucleic acid binding (NAB) Nanoseps have a unique dual layer glass fiber membrane allowing isolation of all nucleic acidsConstructed of low-binding polypropyleneUltrasonically welded seals prevent bypass or seal failureFits standard centrifuge rotors that accept 1.5 mL tubes This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-197-9569