Learn More
Thermo Scientific nanoViper Fingertight Fittings | | 1 EA
Supplier:  Thermo Scientific 6041.5120
Encompass
- Ferrule-free tightening design for virtually zero-dead-volume connections- Works with operating pressures up to 1200 bar- Suitable for temperatures up to 120C- Fingertight design- Flexible PEEK sheathed fused silica capillaries 1/32 in OD- No tools required- Eliminates assembly of PEEK sleeve fittings- Compatible with valves and columns from virtually any manufacturer- Fits narrow connections such as 10-port valves- Available in different materials, lengths, and inner diameters
Catalog No. 50-134-7898