Learn More
Medchemexpress LLC NAP-2/CXCL7 Rat CH 10ug
SDP
Supplier:  Medchemexpress LLC HYP730110UG
Encompass_Preferred
CXCL7 (also known as neutrophil activating peptide 2 NAP-2) is a platelet-derived growth factor that belongs to the ELR CXC chemokine family functioning as a potent chemoattractant and activator of neutrophils through binding to its receptor CXCR2[1] NAP-2/CXCL7 Protein Rat (CHO) is produced in CHO cells and consists of 62 amino acids (I46-I107)
Catalog No. 50-001-80771