Learn More
Medchemexpress LLC Naphazoline | 835-31-4 | 99.7% | 210.27 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY111326100MG
Encompass_Preferred
Naphazoline is a potent α-adrenergic receptor agonist that reduces vascular hyperpermeability and promotes vasoconstriction. It also reduces the levels of inflammatory factors (TNF-α, IL-1β and IL-6), cytokines (IFN-γ and IL-4), IgE, GMCSF, and NGF. It can be used for non-bacterial conjunctivitis research.
  • Reduces vascular hyperpermeability.
  • Promotes vasoconstriction.
  • Reduces inflammatory factors.
  • Reduces cytokines.
  • Reduces IgE, GMCSF, and NGF.
  • Suitable for non-bacterial conjunctivitis research.
Catalog No. 50-003-43340