Learn More
Apexbio Technology LLC Naphazoline HCl 550-99-2 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B13405.1
Encompass_Preferred
Naphazoline HCl (CAS 550-99-2) is a synthetic imidazoline derivative that acts as a sympathomimetic amine It primarily targets -adrenergic receptors on vascular smooth muscle inducing vasoconstriction through the activation of these receptors This property leads to reduced blood flow and decreased vascular congestion particularly in ocular tissues Naphazoline HCl is frequently employed in research to investigate adrenergic signaling pathways and to model vasoconstrictive mechanisms in vitro and in vivo Its pharmacological profile makes it a valuable tool for studies involving vascular responses and ocular pharmacodynamics
Catalog No. 50-000-92535