Learn More
Sigma Aldrich Fine Chemicals Biosciences Naphhol AS-MX phosphae p500MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences N4875500MG
Encompass
Naphthol AS-MX phosphate is soluble in dioxane and is a substrate for phosphatase.
Catalog No. 50-289-7264